Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tie1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Tie1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2384
![]() ![]() |
Proteintech Group Inc
193291AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Probable protein tyrosine-kinase transmembrane receptor.Specifications
Tie1 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P35590, Q06806 | |
21846, 7075, 89806 | |
Tie1 Fusion Protein Ag4728 | |
Primary | |
-20°C | |
TIE1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
JTK14, TIE, TIE1 | |
TIE1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title