Learn More
Tie2 (CD202b) Mouse anti-Human, Mouse, Alexa Fluor 647, Clone: 33 (also known as MoAb33; Ab33), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | Tie2 (CD202b) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 33 (also known as MoAb33; Ab33) |
| Conjugate | Alexa Fluor 647 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | CD202B; TIE-2; TIE2; GLC3E; TEK; TEK tyrosine kinase, endothelial; VMCM; VMCM1 |
| Host Species | Mouse |
| Immunogen | Human TIE2 extracellular domain Recombinant Protein |
| Quantity | 0.1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.