Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIF1 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB1002597
Description
TIF1 alpha Polyclonal specifically detects TIF1 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
TIF1 alpha | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
Rabbit | |
117 kDa | |
100 μL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Naked mole rat, Small-eared galago and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-5 μg/mg lysate, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
O15164 | |
TRIM24 | |
The immunogen recognized by this antibody maps to a region between residue 1,000 and the C-terminus (residue 1050) of human Transcriptional Intermediary Factor 1 Alpha (NP_056989.2). | |
Affinity Purified | |
RUO | |
8805 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction