Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIF1 alpha Antibody (OTI2D9), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274514JF646
Description
TIF1 alpha Monoclonal specifically detects TIF1 alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TIF1 alpha | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRIM24 | |
Recombinant protein fragment corresponding to amino acids 706-1016 of human TIF1 alpha (NP_003843) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D9 | |
Western Blot, Immunohistochemistry | |
E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
Mouse | |
Affinity Purified | |
Protein Kinase, Zinc Finger | |
8805.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction