Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIF1 alpha Antibody (PCRP-TRIM24-1B12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308786AF750
Description
TIF1 alpha Monoclonal antibody specifically detects TIF1 alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
TIF1 alpha | |
Monoclonal | |
Alexa Fluor 750 | |
E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
Recombinant full-length human TIF1 alpha protein (Uniprot: O15164) | |
100 μg | |
Protein Kinase, Zinc Finger | |
8805 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-TRIM24-1B12 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction