Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIF1 alpha Antibody (PCRP-TRIM24-1B12), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308786APC
Description
TIF1 alpha Monoclonal antibody specifically detects TIF1 alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
| TIF1 alpha | |
| Monoclonal | |
| APC | |
| E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
| Recombinant full-length human TIF1 alpha protein (Uniprot: O15164) | |
| 100 μg | |
| Protein Kinase, Zinc Finger | |
| 8805 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-TRIM24-1B12 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction