Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIF1 gamma Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB149922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB149922 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB149922 Supplier Novus Biologicals Supplier No. NBP298000AF350
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TIF1 gamma Polyclonal specifically detects TIF1 gamma in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TIF1 gamma
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase TRIM33, EC 6.3.2, EC 6.3.2.-, Ectodermin homolog, KIAA1113, Protein Rfg7, PTC7FLJ32925, RET-fused gene 7 protein, RFG7FLJ11429, TF1G, TIF1-gamma, TIF1GAMMA, TIF1GECTO, TIFGAMMA, Transcription intermediary factor 1-gamma, transcriptional intermediary factor 1 gamma, tripartite motif containing 33, tripartite motif-containing 33, Tripartite motif-containing protein 33
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TIF1 gamma fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51592
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.