Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIF1 gamma Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281762G
This item is not returnable.
View return policy
Description
TIF1 gamma Polyclonal antibody specifically detects TIF1 gamma in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| TIF1 gamma | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 51592 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| E3 ubiquitin-protein ligase TRIM33, EC 6.3.2, EC 6.3.2.-, Ectodermin homolog, KIAA1113, Protein Rfg7, PTC7FLJ32925, RET-fused gene 7 protein, RFG7FLJ11429, TF1G, TIF1-gamma, TIF1GAMMA, TIF1GECTO, TIFGAMMA, Transcription intermediary factor 1-gamma, transcriptional intermediary factor 1 gamma, tripartite motif containing 33, tripartite motif-containing 33, Tripartite motif-containing protein 33 | |
| TIF1 gamma antibody was raised against an 18 amino acid peptide near the center of human TIF1 gamma. The immunogen is located within amino acids 580 - 630 of TIF1 gamma. Amino Acid Squence: AQNAARIPGIPRHSGPQY | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction