Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIF1 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578496 Supplier Invitrogen™ Supplier No. PA578496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: MCF-7, MDA-MB-231 Predicted Reactivity: Mouse (97%), Xenopus laevis (91%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TIF1 gamma is a member of the Transcriptional Intermediary Factor 1 family, which serve as coactivators and corepressors. Its structure is similar to TIF1 alpha and TIF1 beta, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TIF1 gamma functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIF1 gamma
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TRIM33
Gene Accession No. Q9UPN9
Gene Alias 8030451N04Rik; AI413936; E3 ubiquitin-protein ligase TRIM33; Ecto; Ectodermin homolog; Kiaa1113; Protein Rfg7; PTC7; RET-fused gene 7 protein; RFG7; RING-type E3 ubiquitin transferase TRIM33; TF1G; TIF 1-g; Tif1g; TIF1GAMMA; TIF1-gamma; TIFGAMMA; Transcription intermediary factor 1-gamma; transcriptional intermediary factor 1 gamma; Trim33; tripartite motif containing 33; tripartite motif protein 33; tripartite motif-containing 33; tripartite motif-containing protein 33
Gene Symbols TRIM33
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Human TIF1 gamma. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.