Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIFA Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983065
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3065 Supplier Bioss Supplier No. BS13669R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adapter protein which mediates the IRAK1 and TRAF6 interaction following IL-1 stimulation, resulting in the downstream activation of NF-kappa-B and AP-1 pathways. Induces the oligomerization and polyubiquitination of TRAF6, which leads to the activation of TAK1 and IKK through a proteasome-independent mechanism.

Specifications

Antigen TIFA
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TIFA
Gene Accession No. Q96CG3
Gene Alias TIFAA
Gene Symbols TIFA
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TIFA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92610
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.