Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIFAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561947 Supplier Invitrogen™ Supplier No. PA561947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLSTVNRVSF SGIQMLVRVE EGTSLEAFVC YFHVSPSPLI YRPEAEETDE WEGISQGQ Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.

TIFAB associates with TIFA (MIM 609028) and inhibitsTIFA-mediated activation of NF-kappa-B (NFKB1; MIM 164011).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIFAB
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIFAB
Gene Accession No. Q6ZNK6
Gene Alias TIFA inhibitor; TIFA like protein; TIFA related protein TIFAB; TIFAB; TIFA-like protein; TIFA-related protein TIFAB; TRAF-interacting protein with FHA domain-containing protein B; TRAF-interacting protein with forkhead-associated domain, family member B
Gene Symbols TIFAB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIFAB. Recombinant protein control fragment (Product #RP-100335).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 497189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.