Learn More
Invitrogen™ TIFAB Polyclonal Antibody

Description
TIFAB Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | TIFAB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TIFAB |
| Gene Accession No. | Q6ZNK6, Q8JZM6 |
| Gene Alias | TIFA inhibitor; TIFA like protein; TIFA related protein TIFAB; TIFAB; TIFA-like protein; TIFA-related protein TIFAB; TRAF-interacting protein with FHA domain-containing protein B; TRAF-interacting protein with forkhead-associated domain, family member B |
| Gene Symbols | TIFAB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.