Learn More
Invitrogen™ TIGAR Polyclonal Antibody

Description
This antibody is not recommended for ICC/IF applications.
Specifications
Specifications
| Antigen | TIGAR |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.05% sodium azide |
| Gene | TIGAR |
| Gene Accession No. | Q8BZA9, Q9NQ88 |
| Gene Alias | 9630033F20Rik; AA793651; AI595337; C12orf5; C27H12orf5; C79710; C85509; FR2BP; Fructose-2,6-bisphosphatase TIGAR; fructose-2,6-bisphosphate 2-phosphatase; hypothetical protein LOC502894; LOC502894; probable fructose-2,6-bisphosphatase TIGAR; TIGAR; TP53 induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator; TP53-induced glycolysis regulatory phosphatase; transactivated by NS3TP2 protein; Trp53 induced glycolysis repulatory phosphatase |
| Gene Symbols | TIGAR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.