Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGD1 Antibody (OTI4G9), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274519C
Description
TIGD1 Monoclonal specifically detects TIGD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TIGD1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TIGD1 | |
Recombinant protein fragment corresponding to amino acids 1-267 of human TIGD1 (NP_663748) produced in E.coli. | |
0.1 mL | |
200765.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4G9 | |
Western Blot, Immunohistochemistry | |
EEYORE, tigger transposable element derived 1, tigger transposable element-derived protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction