Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIGD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104178 Supplier Invitrogen™ Supplier No. PA5104178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TIGD2.

TIGD2 belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGD2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIGD2
Gene Accession No. Q0VBL1, Q4W5G0
Gene Alias 3632410O17Rik; epididymis luminal protein 106; HEL106; LOC100912924; Tigd2; tigger transposable element derived 2; tigger transposable element derived 2 homolog; tigger transposable element-derived protein 2; uncharacterized LOC100912924
Gene Symbols TIGD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIGD2(Accession Q4W5G0), corresponding to amino acid residues P272-L322.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912924, 166815, 68140
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.