Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIGD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144061
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144061 Supplier Invitrogen™ Supplier No. PA5144061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human T-47D whole cell, human U-87MG whole cell, human A549 whole cell, human MCF-7 whole cell, rat testis tissue. Flow: Daudi cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The TIGD6 gene encodes a protein belonging to the tigger transposable element derived (TIGD) family, which is a group of proteins that originated from transposable elements but have since evolved to perform various cellular functions. These proteins are implicated in processes such as genomic regulation, chromatin remodeling, and potentially influencing gene expression patterns in eukaryotic cells. TIGD6, in particular, may play a role in cellular differentiation and development, although its specific functions are not yet fully understood. Being part of the tigger-derived family, TIGD6 has connections to genome stability and evolutionary adaptations, influencing how cells respond to developmental cues and maintain genomic integrity. Current research on TIGD6 examines its expression patterns and potential roles in different tissues, particularly regarding implications for cellular adaptation and disease susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGD6
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene TIGD6
Gene Accession No. Q17RP2
Gene Alias TIGD6; tigger transposable element derived 6; tigger transposable element-derived protein 6
Gene Symbols TIGD6
Host Species Rabbit
Immunogen E.coli-derived human TIGD6 recombinant protein (Position: E16-D511).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81789
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.