Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIGD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559532
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559532 Supplier Invitrogen™ Supplier No. PA559532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNKRGKYTTL NLEEKMKVLS RIEAGRSLKS VMDEFGISKS TFYDIKKNKK LILDFVLKQD MPLVGAEKRK RTTGAKYGDV DDAVYMWYQQ KRSAG Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 34%.

The protein encoded by this gene belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGD7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TIGD7
Gene Accession No. Q6NT04
Gene Alias Sancho; TIGD7; tigger transposable element derived 7; tigger transposable element-derived protein 7
Gene Symbols TIGD7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TIGD7. Recombinant protein control fragment (Product #RP-98735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.