Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Tight-Head UN Rated 5 Gal. Carbon Steel Drum
SDP

Catalog No. p-200089213
Click to view available options
Product Type:
Lined Drum
Unlined Drum

Tough, UN Rated drums that are made for hazardous materials.

  • UN Rated and tested to help ensure safe shipping of hazardous materials
  • Tight-head design is equipped with 2 in. NPT and 3/4 in. NPT bungs
  • Choose optional phenolic/epoxy lining for added corrosion resistance
  • Steel lid, body and bottom meet strict wall thickness requirements to ensure strength, durability and long life

Specifications

Product Type Lined Drum
Capacity (English) 55 gal.
Capacity (Metric) 208.2 L
Color Black
Material Carbon Steel
Height (English) 35.25 in.
Height (Metric) 90 cm
Diameter (English) Inner 22.5 in.
Diameter (English) Opening 57.15 cm
Diameter (English) Outer 23 in.
Diameter (Metric) Inner 58 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.