Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tight Junction Protein 2 Antibody, Novus Biologicals™

Guinea Pig Polyclonal Antibody
Supplier: Novus Biologicals NBP157902
Description
Tight Junction Protein 2 Polyclonal specifically detects Tight Junction Protein 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Tight Junction Protein 2 | |
Polyclonal | |
Unconjugated | |
0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide | |
TJP2 | |
For immunisation of the guinea pig, amino acids 411-467 of human ZO-2 were used as immunogen. The antibody cross reacts with mouse ZO-2. | |
0.05 mg | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
0.1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
C9DUPq21.11, DFNA51, DUP9q21.11, MGC26306, Tight junction protein 2, tight junction protein 2 (zona occludens 2), TJP2, X104, X104tight junction protein ZO-2, ZO2, ZO-2, ZO2Friedreich ataxia region gene X104 (tight junction protein ZO-2), Zona occludens protein 2, Zonula occludens protein 2 | |
Guinea Pig | |
IgG purified | |
RUO | |
9414 | |
Store at 4°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction