Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT Antibody (2629A) [Allophycocyanin], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10532A
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Cynomolgus Monkey samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
TIGIT | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Cynomolgus Monkey | |
Purified |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
2629A | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
Human embryonic kidney cell HEK293-derived Cynomolgus monkey TIGIT, Met22-Pro142, Accession # XP_00554815 | |
0.1 mL | |
Cancer, Immunology, Virology Bacteria and Parasites | |
201633 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction