Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT Antibody (741182) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7898J
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
TIGIT | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
741182 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
Mouse myeloma cell line NS0-derived recombinant human TIGIT, Met1-Pro141, Accession # Q495A1 | |
0.1 mL | |
Cancer, Immunology, Virology Bacteria and Parasites | |
201633 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction