Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT Antibody (TIGIT/3106), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279928IR
Description
TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIGIT | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TIGIT | |
Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) (Uniprot: Q4951) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
TIGIT/3106 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction