Learn More
TIGIT Monoclonal Antibody (MBSA43), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$3260.00 - $9860.00
Specifications
Antigen | TIGIT |
---|---|
Clone | MBSA43 |
Concentration | 1 mg/mL |
Classification | Monoclonal |
Conjugate | Functional Grade |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-950-038
![]() |
LIFE TECHNOLOGIES
16950038 |
5 mg |
Each for $3,260.00
|
|
|||||
16-950-0025
![]() |
LIFE TECHNOLOGIES
169500025 |
25 mg |
Each for $9,860.00
|
|
|||||
Description
The MBSA43 MAb reacts with human T cell immunoreceptor with Ig and ITIM domains (TIGIT), which is also known as VSTM3 or WUCAM. This 26 kDa protein is a member of the poliovirus receptor (PVR) family. The expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but it is not expressed on B cells or naive CD4+ T cells. TIGIT may be upregulated on naive CD4+ T cells upon activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK and T cells with antigen presenting cells, fibroblasts, and endothelial cells that express PVR and PVR-like proteins. MBSA43 is reported to crossreact with Rhesus macaque.
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.Specifications
TIGIT | |
1 mg/mL | |
Functional Grade | |
Mouse | |
Human | |
Q495A1 | |
IgG1 κ | |
Affinity Chromatography | |
Antibody |
MBSA43 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
201633 | |
Primary | |
4°C | |
TIGIT |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.