Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT Monoclonal Antibody (MBSA43), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$199.00 - $443.00
Specifications
Antigen | TIGIT |
---|---|
Clone | MBSA43 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H011T03R03A
![]() |
Invitrogen™
H011T03R03A |
100 Tests |
Each for $443.00
|
|
|||||
H011T02R03A
![]() |
Invitrogen™
H011T02R03A |
25 Tests |
Each for $199.00
|
|
|||||
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow CytometrySpecifications
TIGIT | |
5 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
Q495A1 | |
201633 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
MBSA43 | |
Flow Cytometry | |
NovaFluor Red 700 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
IgG1 κ | |
Affinity chromatography | |
TIGIT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title