Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT Mouse anti-Human, BUV395, Clone: 741182, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747845
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow Cytometry.
Specifications
TIGIT | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
TIGIT | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
741182 | |
Brilliant Ultraviolet 395 | |
50 μg | |
Primary | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction