Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIGIT Mouse anti-Human, Clone: 20A9, GenScript™

Catalog No. 502120702
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0702 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0702 Supplier Genscript Corporation Supplier No. A0198140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/mL. T cell immunoreceptor with Ig and ITIM domains (TIGIT), also known as VSTM3, is a type I transmembrane protein, which belongs to the PVR family of immunoglobulin-like domain containing proteins. It is composed of an immunoglobulin variable domain, a transmembrane domain and an immunoreceptor tyrosine-based inhibitory motif (ITIM). TIGIT is expressed on activated T cells, follicular T helper, memory T cells, and regulatory T cells and NK cells. TIGIT binds to CD155 (PVR) with high affinity and it regulates T-cell mediated immunity via the CD226/TIGIT-PVR pathway. TIGIT also serves as a negative regulator of NK and T cell activation.

Human TIGIT Antibody (20A9), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with TIGIT.

TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for ELISA, Flow Cytometry

Specifications

Antigen TIGIT
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 20A9
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene TIGIT
Gene Accession No. Q495A1
Gene Alias ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM
Host Species Mouse
Immunogen Recombinant TIGIT protein
Purification Method Protein A
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201633
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.