Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT, Mouse anti-Human, Clone: TIGIT/3017, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27979320UG
Description
TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
TIGIT | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 1-2 ug/million cells, ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
Mouse | |
Protein A or G purified | |
RUO | |
201633 | |
Store at 4°C. | |
IgG2c κ |
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
TIGIT/3017 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
TIGIT | |
Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction