Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse TIGIT Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB72671S100UG
This item is not returnable.
View return policy
Description
TIGIT Monoclonal specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry.Specifications
TIGIT | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse TIGIT Met1-Thr143 Accession # P86176 | |
100 μg | |
201633 | |
Mouse | |
Supernatant |
Flow Cytometry | |
2190A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P86176 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects mouse TIGIT in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction