Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIGIT Recombinant Mouse Monoclonal Antibody (MBSA43), Alexa Fluor™ Plus 594, Invitrogen™
Mouse Recombinant Monoclonal Antibody
$220.00 - $329.65
Specifications
Antigen | TIGIT |
---|---|
Clone | MBSA43 |
Concentration | 1.0 mg/mL |
Applications | Flow Cytometry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-252-9355
![]() |
Invitrogen™
740010MP59420UG |
20 μg |
Each for $220.00
|
|
|||||
74-001-0MP594
![]() |
Invitrogen™
740010MP594 |
50 μg |
Each
for $329.65
Save 25% Reg : |
|
|||||
Description
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.Specifications
TIGIT | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5 | |
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
TIGIT | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
MBSA43 | |
Flow Cytometry | |
Alexa Fluor Plus 594 | |
Mouse | |
Human | |
Q495A1 | |
201633 | |
IgG1 κ | |
Protein A/G | |
TIGIT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title