Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tildrakizumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541986
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541986 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541986 Supplier Invitrogen™ Supplier No. MA541986
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Tildrakizumab is a high-affinity, humanized, IgG1 kappa antibody targeting interleukin 23 p19 that shows promise in the evolution of treatment strategy in chronic plaque psoriasis 3. The Food and pharmaceutical Administration (FDA) approved ILUMYA (tildrakizumab-asmn) for the treatment of adults with moderate-to-severe plaque psoriasis who are candidates for systemic therapy or phototherapy in March 2018. The approved recommended dosage of ILUMYA is a subcutaneous injection of 100 mg at Weeks 0, 4, and every 12 weeks thereafter 7. A study was performed on the pharmacokinetics of this pharmaceutical on various ethnicities. The pharmacokinetics of tildrakizumab were similar in Japanese, Caucasian, and Chinese subjects 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tildrakizumab Humanized
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias SCH900222; MK-3222
Host Species Human
Immunogen Human IL23A.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.