Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tilfrinib
GREENER_CHOICE

Catalog No. 557910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-791-0 10 mg
55-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 55-791-0 Supplier Tocris Bioscience™ Supplier No. 5579/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective breast tumor kinase (Brk) inhibitor

Tilfrinib is a potent and selective breast tumor kinase (Brk, also known as tyrosine-protein kinase 6, PTK6) inhibitor (IC50 = 3.15 nM). Exhibits >1000-fold selectivity for Brk over a panel of other kinases. Inhibits proliferation of breast cancer cells in vitro.

Specifications

Chemical Name or Material 3-(9H-Pyrido[2,3-b]indol-4-ylamino)phenol
CAS 1600515-49-8
Quantity 10 mg
Target Other Kinase Inhibitors
Molecular Formula C17H13N3O
Formula Weight 275.3
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.