Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Tilted Biohazard Bag Box
SDP

Benchtop biohazard bag holders allow for a cleanable mount for your biohazard bags.

$113.50 - $137.80

Specifications

Product Type Bag Rack
Array 1 x 1
Autoclavable Not Autoclavable
Color Clear
Holds Bags
View More Specs

Products 2
Catalog Number Mfr. No. Height (Metric) Length (Metric) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Height (Metric) Length (Metric) Width (Metric) Price Quantity & Availability  
22-000-796
SDP
View Documents
Poltex™
BIOHTLT8
228 mm 233 mm 89 mm
Each for $113.50
Only null left
Add to Cart
 
22-000-809
SDP
View Documents
Poltex™
BIOHTLT13
381 mm 386 mm 152 mm
Each for $137.80
Only null left
Add to Cart
 
Description

Description

  • Tilted toward the user for better ergonomics in BSC or other tight space
  • Comes in two sizes to fit either 8 x 12 in. or 14 x 19 in. biohazard bags
  • Constructed from super durable and cleanable PETG
  • Rubber bumpons on base for non-slip surface
Specifications

Specifications

Bag Rack
Not Autoclavable
Bags
1
None
Non-sterile
1 x 1
Clear
PETG
Rectangular
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.