Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-1/KIM-1/HAVCR Antibody (104), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290831AF350
Description
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Rat samples. It is validated for ELISA.Specifications
TIM-1/KIM-1/HAVCR | |
Monoclonal | |
Alexa Fluor 350 | |
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat TIM-1/KIM-1/HAVCR (aa18-238; NP_775172.1). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Mouse KIM-1/ TIM1 Human KIM-1/ TIM1 Human TIMD3 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
104 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
26762 | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction