Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-1/KIM-1/HAVCR Antibody (990605), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB17503W
Description
TIM-1/KIM-1/HAVCR Monoclonal antibody specifically detects TIM-1/KIM-1/HAVCR in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
TIM-1/KIM-1/HAVCR | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
990605 | |
Flow Cytometry, CyTOF-ready | |
HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
Human embryonic kidney cell, HEK293-derived human TIM-1/KIM-1/HAVCR, Met1-Thr288, Accession # Q96D42 | |
0.1 mL | |
CD Markers, Cellular Signaling, Immunology | |
26762 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction