Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TIM-1 Monoclonal Antibody (12B1C8H3), Invitrogen™

Catalog No. PIMA543383
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543383 50 μL
PIMA543014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543383 Supplier Invitrogen™ Supplier No. MA543383
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human HAVCR1/TIM1/KIM1

CD365 also known as T-cell immunoglobulin and mucin domain 1 (Tim-1) is a type 1 surface glycoprotein that consists of extracellular N-terminal immunoglobulin V-like domain and a mucin domain, and a short cytoplasmic tail. The endogenous ligand for CD365 is Tim-4. In addition, CD365 is known to be a cellular receptor or co-receptor for a number of viruses including hepatitis A and C viruses, Ebola virus, Marburg virus, Dengue virus, Zika virus, West Nile virus, and Japanese encephalitis virus. The direct binding of the viruses by CD365 occurs via phosphatildyl-serine exposed on viral envelopes. Polymorphisms within human CD365 gene (HAVCR1) are associated with susceptibility to infection and the disease severity. CD365 has also been implicated in the T cell co-stimulation. CD365 is primarily expressed in epithelial cells. It is also highly expressed in A549, 769-P and Caco-2 and cell lines. CD365 exists also as a soluble protein generated by metalloproteinase mediated proteolytic cleavage of the surface protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-1
Applications ELISA
Classification Monoclonal
Clone 12B1C8H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Havcr1
Gene Accession No. Q96D42
Gene Alias AI503787; CD365; HAVCR; HAVCR1; HAVcr-1; Hepatitis A virus cellular receptor 1; hepatitis A virus cellular receptor 1 homolog; Kidney injury molecule 1; Kim1; KIM-1; T cell immunoglobin domain and mucin domain protein 1; t cell immunoglobulin and mucin domain-containing protein 1; T cell membrane protein 1; T-cell immunoglobulin and mucin domain containing 1; T-cell immunoglobulin and mucin domain-containing protein 1; T-cell immunoglobulin mucin family member 1; T-cell immunoglobulin mucin receptor 1; T-cell membrane protein 1; TIM; TIM1; TIM-1; TIMD1; TIMD-1
Gene Symbols Havcr1
Host Species Mouse
Immunogen Recombinant Human HAVCR1 protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26762
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.