Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ TIM-1, Rat, Unlabeled, Clone: 3B3,

Catalog No. MABF245 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF245 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF245 Supplier MilliporeSigma™ Supplier No. MABF245
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Hepatitis A virus cellular receptor 1 homolog (UniProt Q5QNS5; also known as HAVcr-1, Kidney injury molecule 1, KIM-1, T cell immunoglobulin and mucin domain-containing protein 1, T-cell immunoglobulin mucin receptor 1, T cell membrane protein 1, TIM-1, TIMD-1) is encoded by the Havcr1 (also known as Kim1, Tim1, Timd1) gene (Gene ID 171283) in murine species. TIM‐1/KIM‐1 is a phosphatidylserine phagocytosis and scavenger receptor expressed on kidney proximal tubule epithelial cells and, at a much lower level, also among a subset of immune cells. Upon kidney injury, TIM‐1 is upregulated more than a thousand fold and assists tissue injury recovery through phagocytosis of apoptotic cells and cellular debris. KIM‐1‐mediated phagocytosis leads to pro‐tolerogenic antigen presentation, which suppresses CD4+ T‐cell proliferation and increases the percentage of regulatory T-cells in an autophagy gene‐dependent manner. Phagocytosis mediated by KIM‐1 in epithelial cells therefore limits inflammation by both removing dead cells and inducing anti‐inflammatory Treg signaling. Human HAVCR1 polymorphisms are linked to various immunological disorders including allergy, asthma, rheumatoid arthritis, atopic dermatitis, and lupus. TIM-1 is produced with a signal peptide (a.a. 1-21) sequence which is removed posttranslationally to yield the mature protein with a large extracellular region (a.a. 21-290 in human and 22-237 in mouse) composed of a V-type Ig-like domain (a.a. 21-121 in human and 22-122 in mouse) and a mucin domain (the rest of the extracellular region), followed by a transmembrane domain (a.a. 238-258), and a cytoplasmic tail (a.a. 312-359 in human and 259-305 in mouse) with tyrosine phosphorylation sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-1
Applications Functional Assay
Classification Monoclonal
Clone 3B3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat IgG2a in buffer containing PBS without preservatives.
Gene Havcr1, Kim1, Tim1, Timd1
Gene Accession No. Q5QNS5
Host Species Rat
Immunogen Recombinant TIM-1 Ig fusion consists of mouse TIM-1 signal and IgV domains linked to the hinge-CH2-CH3 domains of mouse IgG2a (Umetsu, S.E., et al. (2005). Nat. Immunol.6(5):447-454).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001160103
Test Specificity Clone 3B3 detected recombinant murine TIM-1, but not TIM-3 or TIM-4, Ig fusion. Clone 3B3 immunostained the surface of TIM-1-transfected, but not untransfected 300.19 mouse pre-B-cells (Umetsu, S.E., et al. (2005). Nat. Immunol.6(5):447-454). Clone 3B3 and clone RMT1-10 (Mfr. No. MABF225) exhibited opposite effects toward T cell antigen responses (Xiao, S., et al. (2007). J. Exp. Med. 204(7):1691-1702).
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.