Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-2 Antibody (RM0134-5B23), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122510AF532
Description
TIM-2 Monoclonal specifically detects TIM-2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TIM-2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0134-5B23 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
T Cell immunoglobulin and mucin domain protein 2, Tim2 | |
Mouse recombinant protein of TIM-2 | |
0.1 mL | |
This antibody does not cross-react with mouse TIM-1 and TIM-3. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction