Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TIM-2 Mouse, anti-Mouse, R718, Clone: RMT2-26, BD OptiBuild™

Catalog No. BDB752261 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752261 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752261 Supplier BD Biosciences Supplier No. 752261
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RMT2-26 monoclonal antibody specifically recognizes T cell immunoglobulin and mucin domain-2 (TIM-2) which is also known as T-cell membrane protein 2, TIMD-2, or Tim2. TIM-2 is an ∼55 kDa type I transmembrane glycoprotein that is encoded by Timd2 (T cell immunoglobulin and mucin domain containing 2) which belongs to the immunoglobulin gene superfamily. TIM-2 contains extracellular IgV and mucin domains and a cytoplasmic region with a conserved tyrosine phosphorylation motif which is involved in transmembrane signaling. A human ortholog for mouse TIM-2 has not been identified. TIM-2 is reportedly expressed on B cells with higher levels on activated B cells and germinal center B cells, type-2 T helper (Th2) cells, oligodendrocytes, and epithelial cells in the kidney and liver. It may serve as a receptor for Semaphorin 4A (Sema4A) or the heavy chain of ferritin (H-ferritin). The RMT2-26 antibody can reportedly block the binding of H-ferritin to TIM-2.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-2
Applications Flow Cytometry
Classification Monoclonal
Clone RMT2-26
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.