Learn More
TIM-3 Antibody (2321C), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB23652MFV450
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| TIM-3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cancer, Immunology, Innate Immunity, Neuroscience | |
| 84868 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 2321C | |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession # Q8TDQ0 | |
| RUO | |
| Primary | |
| Human | |
| IgG |