Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 Antibody (2C12), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311989DL594
Description
TIM-3 Monoclonal antibody specifically detects TIM-3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
TIM-3 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG κ |
Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
2C12 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
This antibody was raised by immunising rats with TIM-3:Ig fusion protein emulsified in complete Freund's adjuvant. | |
0.1 mL | |
Immunology, Innate Immunity | |
84868 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction