Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 Antibody (F38-2E2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227221C
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
TIM-3 | |
Monoclonal | |
DyLight 650 | |
Q8TDQ0 | |
HAVCR2 | |
This antibody was raised against recombinant human TIM-3. [Uniprot# Q8TDQ0] | |
0.1 mL | |
Cancer, Immunology, Innate Immunity, Neuroscience | |
84868 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
F38-2E2 | |
Flow Cytometry, Flow (Cell Surface) | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction