Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 Antibody (OTI2E2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274520
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TIM-3 | |
Monoclonal | |
LYOPH | |
Western Blot, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
Mouse | |
33.2 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI2E2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
HAVCR2 | |
Human recombinant protein fragment corresponding to amino acids 22-202 of human HAVCR2(NP_116171) produced in E.coli. | |
Affinity Purified | |
Cancer, Immunology, Innate Immunity, Neuroscience | |
84868 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction