Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, BV711, Clone: 7D3, BD Horizon™
Mouse Monoclonal Antibody
$237.00 - $470.00
Specifications
| Antigen | TIM-3 (CD366) |
|---|---|
| Clone | 7D3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 711 |
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.Specifications
| TIM-3 (CD366) | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human TIM-3 | |
| Primary | |
| Vol. Per test: 5μL |
| 7D3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV711 under optimum conditions and unconjugated antibody and free BD Horizon™ BV711 were removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title