Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 Antibody (OTI2E2), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274520AF647
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for Immunohistochemistry.Specifications
TIM-3 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HAVCR2 | |
Recombinant protein fragment corresponding to amino acids 22-202 of human TIM-3 (NP_116171) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI2E2 | |
Immunohistochemistry | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
Mouse | |
Affinity Purified | |
Cancer, Immunology, Innate Immunity, Neuroscience | |
84868.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction