Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3, Mouse anti-Human, Clone: TIM3/3113, Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $536.00
Specifications
Antigen | TIM-3 |
---|---|
Clone | TIM3/3113 |
Concentration | 0.2 mg/mL |
Dilution | ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
TIM-3 | |
0.2 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Immunology, Innate Immunity | |
10 mM PBS with 0.05% BSA | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
HAVCR2 | |
IgG2a κ | |
Protein A or G purified |
TIM3/3113 | |
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q8TDQ0, Q8TDQ0 | |
84868 | |
Recombinant fragment of human TIM-3 protein (around aa 22-202) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title