Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3 Rabbit anti-Human, Clone: 24, Novus Biologicals™

Rabbit Monoclonal Antibody
$402.00 - $572.50
Specifications
Antigen | TIM-3 |
---|---|
Clone | 24 |
Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for ELISA, Sandwich ELISA Capture.Specifications
TIM-3 | |
ELISA 1:5000-1:10000, Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
Recombinant Human TIM-3 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
24 | |
ELISA | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity | |
0.2 um filtered solution in PBS | |
84868 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title