Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-3, Rat anti-Mouse, Clone: TI 142F, Novus Biologicals™

Rat Monoclonal Antibody
$342.00 - $464.00
Specifications
Antigen | TIM-3 |
---|---|
Clone | TI 142F |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, ELISA |
Applications | Flow Cytometry, ELISA |
Description
TIM-3 Monoclonal specifically detects TIM-3 in Mouse samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.Specifications
TIM-3 | |
1 mg/mL | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rat | |
Immunology, Innate Immunity | |
0.2 um-filtered solution in PBS, pH 7.4 | |
84868 | |
Recombinant mouse TIM-3. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
TI 142F | |
Flow Cytometry, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
HAVCR2 | |
IgG2a κ | |
Protein G purified | |
Recognizes mouse TIM-3. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title