Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-4 Antibody (102), DyLight 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290556V
Description
TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for ELISA.Specifications
TIM-4 | |
Monoclonal | |
DyLight 405 | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TIM-4 (NP_848874.3; Met 1-Thr 279). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human TIMD4 Human KIM1 (aa 1-135) | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
102 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
91937 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction