Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-4 Antibody (3A1), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311990PE
Description
TIM-4 Monoclonal antibody specifically detects TIM-4 in Mouse samples. It is validated for Flow CytometrySpecifications
TIM-4 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
91937 | |
Store at 4°C in the dark. |
Flow Cytometry | |
3A1 | |
Flow Cytometry | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 | |
This antibody was raised by immunising female Lewis rats (Harlan Sprague-Dawley) with TIM-4:Ig fusion protein in complete Freund's adjuvant. | |
0.1 mL | |
Primary | |
Mouse | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction