Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIM-4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176702
Description
TIM-4 Polyclonal specifically detects TIM-4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry.Specifications
TIM-4 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 | |
Rabbit | |
45 kDa | |
0.1 mg | |
Primary | |
At least four isoforms of TIM-4 are known to exist; this antibody will detect all four isoforms. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunocytochemistry | |
1 mg/mL | |
Western Blot 2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 10-20 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml | |
Q96H15 | |
TIMD4 | |
Antibody was raised against a 16 amino acid synthetic peptide from near the center of human TIM-4. The immunogen is located within amino acids 60 - 110 of TIM-4. Amino Acid Squence: RTDGMRVTSRKSAKYR | |
Affinity Purified | |
RUO | |
91937 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction